Orders LinkContactUsLinkComponent
Safety helmets, 1100
Safety helmets, 1100
Safety helmets, 1100
Certificates
Safety helmets, 1100
Safety helmets, 1100
4 Options Available Below
Product Details & Documents
Protective helmet with helmet shell made from high density polyethylene (HDPE). For use in construction and industry.
  • With six point LDPE inner cradle
  • Ear protection, face protection shield and chin strap available as accessories
Size: 510 to 630 mm
Weight: 375 g
EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)
Certifications: EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)
Documents
Product Family Options
Without ratchet
Product Information
  • Colour
    Availability
    Price
  • 111-1502
    Safety helmet, unvented, UK
    Safety helmet, unvented, UK
    Blue
    Discontinued
  • 111-1512
    Safety helmet, unvented, UK
    Safety helmet, unvented, UK
    White
    Discontinued
  • 111-1513
    Safety helmet, unvented, UK
    Safety helmet, unvented, UK
    Yellow
    Discontinued
  • 111-1616
    Safety helmet, 1100, unvented, with full peak and sweatband
    Safety helmet, 1100, unvented, with full peak and sweatband
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,70 €
    10,70 €
    1 * 1 ST
       10,70 €
  • 111-0854
    Safety helmet, 1100, unvented, with full peak and sweatband
    Safety helmet, 1100, unvented, with full peak and sweatband
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
With ratchet
Product Information
  • Colour
    Availability
    Price
  • 111-1675
    Safety helmet, 1100, unvented, with full peak, with ratchet
    Safety helmet, 1100, unvented, with full peak, with ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • CENUS01EYRC
    Safety helmet, 1100, unvented, with full peak, with ratchet
    Safety helmet, 1100, unvented, with full peak, with ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST