Closing cones, Combi-Stoppers
Supplier: B. Braun
Jump to:
About this item
These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
- Unique multifunctional design
- Sterile closing of pre-filled syringes and all kinds of open IV-accesses in IV-Sets
Available in red, blue and white.
3 Options Available Below
Product Details & Documents
These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
- Unique multifunctional design
- Sterile closing of pre-filled syringes and all kinds of open IV-accesses in IV-Sets
Available in red, blue and white.
Documents
Product Family Options
Product Information
- ColourAvailabilityPrice
- BRAU4495152Closing cones for needle-free injection sites and seal pre-filled syringesClosing cones for needle-free injection sites and seal pre-filled syringesBlueLog in for availability1 * 100 ST53,80 €
- 612-0100Closing cones for needle-free injection sites and seal pre-filled syringesClosing cones for needle-free injection sites and seal pre-filled syringesRedLog in for availability1 * 100 ST53,80 €
- BRAU4495209Closing cones for needle-free injection sites and seal pre-filled syringesClosing cones for needle-free injection sites and seal pre-filled syringesWhiteLog in for availability1 * 100 ST53,80 €