Order Entry
Belgium
ContactUsLinkComponent
Burner, Teclu, with stopcock
Burner, Teclu, with stopcock
Catalog # USBE1240
undefined
Burner, Teclu, with stopcock
Catalog # USBE1240

Specifications

  • Max. temp.:
    1620 °C
  • Description:
    Teclu burner with lever stopcock, pilot flame and air regulation
  • Head Ø:
    17 mm
  • Pk:
    1
  • For:
    Natural gas

Specifications

About this item

Burner for natural gas, adjustable. Maximum temperature of 1620 °C.