Sand, crystalline Ø 0.1 - 0.5 mm, washed, calcinated
Fournisseur: GERBER FUNKE
Certificats
À propos de cet article
Numéro CAS:7631-86-9
:14808-60-7
Formule:SiO₂
Numéro MDL:MFCD02100519
:60.08 g/mol
Frequently Bought Together
Potassium dihydrogen phosphate, EMSURE® ISO for analysis, Supelco®
Fournisseur: Merck
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Fournisseur: VWR Chemicals
VWR® Lisbon, Visitor Spectacles
Fournisseur: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Fournisseur: VWR Chemicals
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Fournisseur: Merck
Potassium chloride, EMSURE® for analysis, Supelco®
Fournisseur: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Fournisseur: Merck
Sodium chloride regeneration salt, TECHNICAL for water softening
Fournisseur: VWR Chemicals
Transport container with screw cap
Fournisseur: BOETTGER PAUL