Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Digital mini surface thermometer

Mini thermometer for surface measurements, −50…+300 °C, ±1,0 (−30…+250);
±2,0 °C, 0,1 (−19,9…+199,9);
1,0 °C, NTC (Lר: 120×15 mm)
Catalog # TEST0560.1109
Supplier:   TESTO
Digital mini surface thermometer
Catalog # TEST0560.1109
Supplier:  TESTO
Digital mini surface thermometer
Digital mini surface thermometer

Mini thermometer for surface measurements, −50…+300 °C, ±1,0 (−30…+250);
±2,0 °C, 0,1 (−19,9…+199,9);
1,0 °C, NTC (Lר: 120×15 mm)

Price
37,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The mini surface thermometer is used for the most varied methods of surface temperature measurements (for temperature checks in heating engineering, the sanitation sector, or air conditioning/refrigeration and ventilation technology).

  • Universally applicable for surface measurement
  • Large, easy-to-read display
  • Battery status display and easy battery replacement
  • Stainless steel surface temperature probe
  • Auto power-off (1 hour) with low battery display
  • Selectable °C/°F display

Silicon thermal paste can be used for fast measurement results which provides optimal thermal transference between the measuring tip and object being measured.

 : Supplied with batteries.

Specifications
  • Range:
    −50…+300 °C
  • Resolution:
    0,1 (−19,9…+199,9);
    1,0 °C
  • Description:
    Mini thermometer for surface measurements
  • Accuracy:
    ±1,0 (−30…+250);
    ±2,0 °C
  • Probe:
    NTC (Lר: 120×15 mm)