Order Entry
Belgium
ContactUsLinkComponent
Bovine Ribonuclease A (from Pancreas)
Bovine Ribonuclease A (from Pancreas)
Certificates
Bovine Ribonuclease A (from Pancreas)

Specifications

  • CAS No.:
    9001-99-4
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00132184
  • Enzyme name:
    Ribonuclease A
  • Enzyme synonyms:
    RNase A

Specifications

Frequently Bought Together