Ribonuclease A
Catalog # ROTH7156.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- CAS No.:9001-99-4
- Storage conditions:Freezer
- MDL No.:MFCD00132184
- Enzyme name:Ribonuclease A
- Enzyme synonyms:RNase A