Order Entry
Belgium
ContactUsLinkComponent
Bovine desoxyribonuclease I (from Pancreas)
Bovine desoxyribonuclease I (from Pancreas)
Supplier:  PanReac AppliChem
Certificates
Bovine desoxyribonuclease I (from Pancreas)
Supplier:  PanReac AppliChem
Certificates

Specifications

  • CAS No.:
    9003-98-9
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Freezer
  • Enzyme name:
    Desoxyribonuclease I
  • Enzyme synonyms:
    Dnase I

Specifications

Related Information

Frequently Bought Together