Order Entry
Belgium
ContactUsLinkComponent
Chicken lysozyme (Muramidase) (from egg white)
Chicken lysozyme (Muramidase) (from egg white)
Catalog # ROCKMB-109-1000
CAS Number:  
Chicken lysozyme (Muramidase) (from egg white)
Catalog # ROCKMB-109-1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • CAS No.:
    12650-88-3
  • Source:
    Egg white
  • Species:
    Chicken
  • Storage conditions:
    Freezer
  • Enzyme name:
    Lysozyme (Muramidase)

Specifications

About this item