Order Entry
Belgium
ContactUsLinkComponent
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer) pH 8,3
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer) pH 8,3
Catalog # A1415.1000
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer) pH 8,3
Catalog # A1415.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer) pH 8,3

Specifications

About this item