Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Supplier: PanReac AppliChem
Certificates
Certificates
About this item
Frequently Bought Together
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF
Magnetic stirrers, MIX series
Supplier: 2MAG
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
Ammonium iron (III) citrate (∼18% Fe), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
Calcium chloride ≥98.0% (dried basis), powder, AnalaR NORMAPUR® Reag. Ph. Eur.
Supplier: VWR Chemicals
Water, Molecular Biology Grade nuclease, DNase-, RNase-free, sterile
Supplier: VWR Chemicals
Measuring scoops
Supplier: BURKLE
Decontamination Solution, DNA/RNA-ExitusPlus™
Supplier: PanReac AppliChem
Kits for Precellys®
Supplier: Bertin Instruments
Microcentrifuge tubes, 1,5 ml
Supplier: KARTELL
VWR® Automatic Burettes According to Dr. Schilling
Supplier: VWR Collection
VWR®, Combs for PerfectBlue™ Gel System Maxi M
Supplier: VWR Collection
VWR®, Accessories for VWR Bead Mill Homogenisers
Supplier: VWR Collection
TOF G2-S SAMPLE KIT -1 1 * 1 KIT
Supplier: WATERS
LIF TEST MIX REFR 1 * 1 items
Supplier: EKSIGENT
ZIP BAG LDPE 400x600x0.09MM 1 * 250 items
Supplier: HK PACK KRAUTSCHEID