Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0, Glass bottle, 100 mL
Catalog # SERA39799.01
Marketsource
Supplier:   Serva
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0
Catalog # SERA39799.01
Supplier:  Serva
Marketsource
CAS Number:  
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0, Glass bottle, 100 mL

CAS Number:  
Price
28,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.