Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L
Catalog # MOLE11225962-1L
Marketsource
Supplier:   Molekula
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0
Catalog # MOLE11225962-1L
Supplier:  Molekula
Marketsource
CAS Number:  
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.