Buffer, TE solution 100x concentrate (TRIS-EDTA buffer), Millipore®
TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 1 L
Supplier: Merck Millipore (Calbiochem)
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer), Millipore®
Catalog # 574793-1L
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:TE buffer solution 100x concentrate (TRIS-EDTA buffer)



