Order Entry
Belgium
ContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l), Millipore®
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l), Millipore®
Catalog # 324504-500
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l), Millipore®
Catalog # 324504-500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications

About this item