Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, PBS solution 10X concentrate (phosphate buffered saline)

Buffer, PBS solution 10X concentrate (phosphate buffered saline)

PBS buffer solution 10X concentrate (phosphate buffered saline), Glass bottle, 1 L
Catalog # ROTH1058.1
Marketsource
Supplier:   Roth Carl
Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # ROTH1058.1
Supplier:  Roth Carl
Marketsource
CAS Number:  
Buffer, PBS solution 10X concentrate (phosphate buffered saline)

PBS buffer solution 10X concentrate (phosphate buffered saline), Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    PBS buffer solution 10X concentrate (phosphate buffered saline)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.