Order Entry
Belgium
ContactUsLinkComponent
VWR® UltraPure, Buffer, concentrate (phosphate buffered saline)
VWR® UltraPure, Buffer, concentrate (phosphate buffered saline)
Catalog # J373-4L
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® UltraPure, Buffer, concentrate (phosphate buffered saline)
Catalog # J373-4L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    4 L
  • Description:
    PBS buffer solution 10X concentrate (phosphate buffered saline)

Specifications

About this item

  • pH at 25 °C (10% in water): 7,3 - 7,5
  • Conductivity (10% in water): 14000 - 17000 mhos
  • No DNases, RNases or proteases detected, sterile