
Ampicillin sodium salt 91.0-102.0%, BioChemica®
:69-52-3
:
:C₁₆H₁₈N₃NaO₄S
:MFCD00064313
:371.39 g/mol
:212…215 °C (dec.)
:200-708-1
:GHS08
:Refrigerator
- Assay (HPLC):91.0 - 102.0 %
- α20°C/D; 0.2 %, H2O:+258° - +287°
- pH (10 %; H2O; 20°C):8.0 - 10.0
- Heavy metals:max. 0.002 %
- Water (K.F.):max. 2 %
Frequently Bought Together