
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98%, Ultra Pure Grade
Catalog # N679-10G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Abs. @300nm (5%, Water):<0.15
- Abs. @400nm (5%, Water):<0.06
- Dioxane Test:Pass
- Appearance:White crystalline powder
- Melting Point:110 - 114 °C
- Moisture (KF):<1%
- pH (5%, Water) @25 °C:5 - 7
- Purity:>98%
- Specific Rotation:-34.5 – -28.5°
Specification Test Results
About this item
:
:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109,5…110,5 °C
EINECS:206-703-0
:GHS07
Storage Temperature:Freezer
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273