Order Entry
Belgium
ContactUsLinkComponent
 
Desiccators, vacuum, with plate, SP Bel-Art
Catalog # BELAF420430000
undefined
Desiccators, vacuum, with plate, SP Bel-Art
Catalog # BELAF420430000

Specifications

  • Type:
    With stopcock
  • Description:
    Vacuum chamber and plate, 5 l
  • Pk:
    1
  • Ø ext.×H:
    228×160 mm

Specifications

About this item

Ideal for vacuum drying, degassing and teaching demonstrations, this vacuum chamber can withstand a full vacuum at room temperature.

  • One piece O-ring seals the clear polycarbonate top to the white polycarbonate base plate
  • Polypropylene stopcock with Teflon® PTFE plug and tapered hose barb accepts tubing with 6 mm int.Ø.