ContactUsLinkComponent
Dial caliper, SP Bel-Art
Dial caliper, SP Bel-Art
Catalog # BELAH134160001
undefined
Dial caliper, SP Bel-Art
Catalog # BELAH134160001
Specifications
  • Range:
    0 − 150 mm
  • Description:
    Dial caliper
  • Pk:
    1
Specifications
About this item
This dial caliper can measure up to 150 mm on a white dial graduated in 0,1 mm increments. Vernier model measures 130 mm in 0,05 subdivisions, plus 5" scale with 0,001 subdivisions.
  • Adjustable dial for zero settings and differential measurements
  • Easy-to-use thumb roller adjusts jaws to measure internal and external diameters, and depth
Designed to measure glassware and plasticware accurately and safely, with durable plastic polymer.