Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Automatic tilt measures

Supplier:   GERBER FUNKE
Automatic tilt measures
Supplier:  GERBER FUNKE
Certificates
Automatic tilt measures
Automatic tilt measures

2 Options Available Below

Product Details & Documents

Glass, with rubber stopper.

  • Superior quality models
  • Storage bottle of 500/250 ml
  • For milk analysis applications

Colour: Clear

Product Family Options

Product Information

  • Capacity
    Colour
    Availability
    Price
  • 614-0148
    Amyl alcohol
    Amyl alcohol
    1 ml
    Clear
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,80 €
    60,80 €
    1 * 1 ST
       60,80 €

  • 614-0147
    Sulphuric acid
    Sulphuric acid
    10 ml
    Clear
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,20 €
    70,20 €
    1 * 1 ST
       70,20 €