Sediment testers for milk
Fournisseur: GERBER FUNKE
Certificats
À propos de cet article
SEDILAB: Manual sediment tester. With clamp for attaching to tables.
SEDILAB-E: Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk. Splash-proof design. Approx. 800 samples per hour. Sharply defined sediment images.
Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk.
Frequently Bought Together