Orders LinkContactUsLinkComponent
Shakers
Shakers
Supplier:   GERBER FUNKE
Shakers
Supplier:  GERBER FUNKE
Certificates
Shakers
3 Options Available Below
Product Details & Documents
Shakers for determining the fat content in milk according to Mojonnier's method. White PVC housing. Choice of models to accept 4, 6 or 36 Mojonnier extraction tubes. Units provide forceful, uniform and reproducible mixing.
  • Timer, 0 to 15 min
  • Produce 100 shakes per minute
  • Model for 36 butyrometers is supplied with stand
Product Family Options
Product Information
  • Availability
    Price
  • 614-0184
    Shaker for six Mojonnier tubes
    Shaker for six Mojonnier tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 390,00 €
    2 390,00 €
    1 * 1 ST
       2 390,00 €
  • 614-0183
    Shaker for four Mojonnier tubes
    Shaker for four Mojonnier tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 340,00 €
    2 340,00 €
    1 * 1 ST
       2 340,00 €
  • 614-0185
    Shaker and stand for thirty six Mojonnier tubes
    Shaker and stand for thirty six Mojonnier tubes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cork stopper for extraction tubes, Mojonnier`s method
    Cork stopper for extraction tubes, Mojonnier`s method
    614-0188
    Log in for availability
    1 * 1 ST - 0,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Extraction tubes, Mojonnier, with flattened bulb
    Extraction tubes, Mojonnier, with flattened bulb
    614-0187
    Log in for availability
    1 * 1 ST - 23,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Extraction tubes, Mojonnier, with rounded bulb
    Extraction tubes, Mojonnier, with rounded bulb
    614-0186
    Log in for availability
    1 * 1 ST - 23,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Stand, wooden, for 12 extraction tubes acc. to Mojonnier`s method
    Stand, wooden, for 12 extraction tubes acc. to Mojonnier`s method
    614-0189
    Log in for availability
    1 * 1 ST - 86,90 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.