ContactUsLinkComponent
Caps for Nunc™ TufRol™ roller bottles
Caps for Nunc™ TufRol™ roller bottles
Catalog # 392-0326
   Low Manufacturing Impact
undefined
Caps for Nunc™ TufRol™ roller bottles
Catalog # 392-0326
   Low Manufacturing Impact
Specifications
  • Description:
    Vented caps for TufRol™ roller bottles, HDPE, sterile
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Pk:
    500
  • Environmental certification:
    NAR Certificate of Retirement
Specifications
About this item
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904