Orders LinkContactUsLinkComponent
EasyReach™ Cleaning Tools
EasyReach™ Cleaning Tools
Supplier:   CONTEC
EasyReach™ Cleaning Tools
Supplier:  CONTEC
Certificates
EasyReach™ Cleaning Tools
EasyReach™ Cleaning Tools
9 Options Available Below
Product Details & Documents
The lightweight, stainless steel EasyReach™ Cleaning Tool is designed for cleaning isolators, laminar air flow hoods, glove boxes, and other mini environments. Rectangular shape makes overlapping straight-line action easy to achieve. Hinged joint and lightweight design makes one-handed cleaning comfortable and improves access to hard-to-reach areas.
  • Rounded tool edges and strong, durable, double-layer pads ensure equipment is not damaged during use
  • Lightweight EasyReach™ Tool can easily be manipulated, is comfortable to use, and minimizes fatigue
  • All components are designed to be chemical resistant, minimizing the risk of contamination from degradation
  • Presaturated option removes the need for 70% IPA spray bottles, reducing inventory and minimizing waste costs
  • Sorbent double layer Quiltec sealed edge polyester cleaning pads ensure particles and fibers are minimized
  • Polyester cleaning pads leave no unwanted surface residues
  • Available validated sterile
The single-use 100% polyester EasyReach™ cleaning pads provide a low level of particulates which is essential for use in product contact areas. The pads are very absorbent, ensuring even coverage of disinfectant or IPA. Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame, but is easy to replace with gloved hands. Sterile pads presaturated with 70% IPA are also available.
Product Family Options
EasyReach™ cleaning pads
Product Information
  • L×W
    Length
    Packed
    Availability
    Price
  • 129-0574
    Cleaning pads, EasyReach™, quilted polyester, sewn
    Cleaning pads, EasyReach™, quilted polyester, sewn
    190×71 mm (7.5×2.8")
    4 each/bag, 15 bags/case
    Log in for availability
    Log in for availability
    1 * 60 ST
    1 * 60 ST - 488,00 €
    488,00 €
    1 * 60 ST
       488,00 €
  • 129-2470
    Cleaning pads, EasyReach™, 100% knitted polyester, sterile
    Cleaning pads, EasyReach™, 100% knitted polyester, sterile
    190×71 mm (7.5×2.8")
    4 each/bag, 15 bags/case
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 60 ST
    1 * 60 ST - 394,00 €
    394,00 €
    Log in for availability
    Each
    Each - 378,00 €
    378,00 €
    1 * 60 ST
       394,00 €
    Each
       378,00 €
  • 129-1655
    Cleaning pads, EasyReach™, quilted polyester, ultrasonically sealed
    Cleaning pads, EasyReach™, quilted polyester, ultrasonically sealed
    190×71 mm (7.5×2.8")
    4 each/bag, 25 bags/case
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 730,00 €
    730,00 €
    1 * 100 ST
       730,00 €
  • 129-1656
    Cleaning pads, EasyReach™, quilted polyester, sterile, ultrasonically sealed
    Cleaning pads, EasyReach™, quilted polyester, sterile, ultrasonically sealed
    190×71 mm (7.5×2.8")
    4 each/bag, 25 bags/case
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 765,00 €
    765,00 €
    1 * 100 ST
       765,00 €
  • 115-2814
    Cleaning pads, EasyReach™, 100% knitted polyester, sterile, presaturated with 70% IPA and 30% DI water
    Cleaning pads, EasyReach™, 100% knitted polyester, sterile, presaturated with 70% IPA and 30% DI water
    190×71 mm (7.5×2.8")
    8 each/bag, 8 bags/case
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 64 ST
    1 * 64 ST - 690,00 €
    690,00 €
    Log in for availability
    Each
    Each - 663,00 €
    663,00 €
    1 * 64 ST
       690,00 €
    Each
       663,00 €
EasyReach™ cleaning tools
Product Information
  • L×W
    Length
    Packed
    Availability
    Price
  • CONT2685P
    QuickConnect™ EasyReach™ lay-flat frame, polypropylene
    QuickConnect™ EasyReach™ lay-flat frame, polypropylene
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,60 €
    19,60 €
    1 * 1 ST
       19,60 €
  • CONT2685LF
    EasyReach™ stainless steel lay flat mop frame
    EasyReach™ stainless steel lay flat mop frame
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 204,00 €
    204,00 €
    1 * 1 ST
       204,00 €
  • CONT2665LF
    EasyReach™ stainless steel one-piece lay flat frame
    EasyReach™ stainless steel one-piece lay flat frame
    420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 210,00 €
    210,00 €
    1 * 1 ST
       210,00 €
  • 129-0204
    QuickConnect handle, telescopic, stainless steel
    QuickConnect handle, telescopic, stainless steel
    400 - 760 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 401,00 €
    401,00 €
    1 * 1 ST
       401,00 €