ContactUsLinkComponent
Working tray, benchtop, SP Bel-Art
Working tray, benchtop, SP Bel-Art
Catalog # BELAF246750060
undefined
Working tray, benchtop, SP Bel-Art
Catalog # BELAF246750060
Specifications
  • Width:
    560 mm
  • Height:
    13 mm
  • Length:
    460 mm
  • Pk:
    1
Specifications
About this item
Made of Polypropylene.
  • Can be used in combination with biohazard, radioactive or general purpose mats
  • Tray lip assures containment of spilled liquids that are not absorbed by the mats
  • Autoclavable
This plastic tray is designed to hold the most popular sizes of safety mats to help contain spills.