VWR®, Rapid Transfer buffer 10X
Rapid transfer buffer, 10X, 4 L
Supplier: VWR Chemicals
VWR®, Rapid Transfer buffer 10X
Catalog # N789-4L
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
Rapid transfer buffer is a simple, single component system for quick and efficient transfer of proteins from SDS-PAGE gels to membranes for Western blotting applications. Transfer is completed in 10 or 20 minutes using a standard semi-dry or wet transfer apparatus, respectively.
- Utilises standard transfer equipment
- Non hazardous formulation
- Transfers to PVDF and nitrocellulose from multiple types of gels
Rapid transfer buffer is a methanol-free, non hazardous formulation that is compatible with PVDF and nitrocellulose membranes. The transfer efficiency is equivalent to that observed when using a Tris-glycine-methanol transfer buffer.
Specifications
Specifications
- Pk:4 L
- Description:Rapid transfer buffer, 10X
- Cat. No.:N789-4L
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.