Magnetic stirring bar retriever, HDPE, SP Bel-Art
Catalog # BELAF377750000
Supplier: Bel-Art Products, a Part of SP
Specifications
- For:External use, avoids contact with the liquid and possible contamination
- Length:200 mm
- Pk:1
Specifications
About this item
This retriever is able to remove a magnetic stirring bar without coming into contact with the contents of the vessel.
- Eliminates contamination of contents
- Does not need to be cleaned or autoclaved after use
- Long ribbed handle provides a secure grip