ContactUsLinkComponent
Magnetic stirring bar retrievers, PTFE, jumbo, Spinbar®, SP Bel-Art
Magnetic stirring bar retrievers, PTFE, jumbo, Spinbar®, SP Bel-Art
Catalog # BELAF377810000
undefined
Magnetic stirring bar retrievers, PTFE, jumbo, Spinbar®, SP Bel-Art
Catalog # BELAF377810000
Specifications
  • Ø:
    100 mm
  • Description:
    With "T" handle
  • Length:
    650 mm
  • Pk:
    1
Specifications
About this item
Flexible PTFE magnetic retriever for removing the largest magnetic stirring bars from deep vessels. Designed with powerful Alnico 5 magnetic end having 102 mm in length.
  • Easy grip 'T' handle
  • Longer, custom lengths are available