Magnetic stirring bars, PTFE, polygonal, assorted, Spinbox™, SP Bel-Art
Catalog # BELAF371840000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Type:Assortment of 18 polygon shaped magnetic stirring bars
- Environmentally Preferable:
- Pk:1 SET
Specifications
About this item
Magnetic stirring bars with Alnico magnet, PTFE encapsulated is supplied as an assortment and comes in a convenient storage box to provide instant availability.
- Low friction PTFE coating offers superior chemical compatibility
Delivery: Assortment consists of 18 assorted bars, each two of the following sizes: 10×6 mm, 15×6 mm, 20×7 mm, 25×8 mm, 30×8 mm, 40×8 mm, 50×8 mm, 60×8 mm and 80×10 mm.