Order Entry
Belgium
ContactUsLinkComponent
 
Magnetic stirring bars, PTFE, for spectrophotometer cells or cuvettes, Spinbar®, SP Bel-Art
Catalog # BELAF371500000
undefined
Magnetic stirring bars, PTFE, for spectrophotometer cells or cuvettes, Spinbar®, SP Bel-Art
Catalog # BELAF371500000

Specifications

  • Ø:
    9 mm
  • For:
    Standard 10 mm light path cuvette
  • Height:
    8 mm
  • Pk:
    1

Specifications

About this item

This magnetic stirring bar, with Alnico magnet encapsulated in PTFE coating, fits into the standard 10 mm light path cuvette and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine.

  • Centrifugal pumping action, generated by the cross channels in the upper face, mixes without aeration
  • Photometric measurements can proceed uninterrupted while reagents are added
  • One-piece moulded unit will not scratch cuvettes; may also be used in test tubes
  • Autoclavable
Certifications: Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification