ContactUsLinkComponent
Pipette tray, SP Bel-Art
Pipette tray, SP Bel-Art
Catalog # BELAF189400000
undefined
Pipette tray, SP Bel-Art
Catalog # BELAF189400000
Specifications
  • For:
    12 short, 16 medium or 7 long pipettes
  • Description:
    Pipette tray
  • W×D×H:
    286×216×29 mm
  • Pk:
    1
Specifications
About this item
This pipette tray made of PS can be used in drawers or on the benchtop for storing pipettes and can also be used for soaking pipettes before cleaning.
  • Scalloped rims hold 16 small or 7 large pipettes
  • Center support holds 12 short pipettes in a perpendicular direction