ContactUsLinkComponent
Ethidium bromide 0,625 mg/l in aqueous solution, Biotechnology Grade
  E406-5ML
 :  
undefined
Ethidium bromide 0,625 mg/l in aqueous solution, Biotechnology Grade
  E406-5ML
 :  
  • Pack type:
    Dropping bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:
  • Abs.@ Lambda Max:
    0.68 – 0.74
  • DNase:
    none detected
  • Lambda Max (8/100 ml Water):
    478 – 482 nm
  • RNase:
    none detected
 : 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
 :
 :394.31 g/mol
 :214-984-6
 :1239-45-8
 :C₂₁H₂₀BrN₃
 :MFCD00011724