ContactUsLinkComponent
VWR®, SSC buffer, 20X, ultra-pure grade
VWR®, SSC buffer, 20X, ultra-pure grade
Catalog # 0804-4L
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, SSC buffer, 20X, ultra-pure grade
Catalog # 0804-4L
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Bag-in-box
  • Pk:
    4 L
  • Description:
    SSC buffer, 20X liquid concentrate
  • Pk Info:
    -
Specifications
About this item
SSC buffer is a hybridisation buffer for Northern and Southern blots.
Delivery: Supplied as either 20X liquid concentrate or Ready-Pack™ that prepares 1 litre of 20X concentrate.