Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides
Supplier:  MARIENFELD
Certificates
Cavity slides
4 Options Available Below
Product Details & Documents
Soda-lime glass of hydrolytic class 3 cavity slides.
Carefully ground and polished for in vitro diagnostic (IVD) applications.
Certifications: IVD 98/79/EC
Documents
Product Family Options
Product Information
  • No. of wells
    Availability
    Price
  • Cavity slides
    1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 25,70 €
    25,70 €
    1 * 10 ST
       25,70 €
  • Cavity slides
    1
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 111,00 €
    111,00 €
    1 * 50 ST
       111,00 €
  • 630-1612
    Cavity slides
    Cavity slides
    2
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 170,00 €
    170,00 €
    1 * 50 ST
       170,00 €
  • 630-1613
    Cavity slides
    Cavity slides
    3
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 231,00 €
    231,00 €
    1 * 50 ST
       231,00 €