VWR®, Protease inhibitor cocktails, general use
Supplier: VWR Chemicals
Certificates
About this item
Contains a mixture of protease inhibitors: AEBSF, Aprotinin, E-64, Bestatin and Leupeptin. Each vial can be reconstituted in 1 ml deionised water to form a 100X solution.
- Effectively preserves the native state of proteins
M222 with EDTA