Tridemorph, Supelco®
Glass bottle for solids, 100 mg
N° de catalogue 36199-100MG
Fournisseur: Merck
Aller à :
Product Details & Documents
- Numéro CAS:24602-86-6
- Formule:C₁₉H₃₉NO
- :M-Clarity™ quality level = MQ100
- EINECS:246-347-3
Spécifications
Spécifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



