Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hydroxyectoine

Hydroxyectoine

Hydroxyectoine
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:165542-15-4
  • Formula:C₆H₁₀N₂O₃
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.