Order Entry
Belgium
Orders LinkContactUsLinkComponent
γ-Cyclodextrine

γ-Cyclodextrine

Glass bottle for solids, 1 g
Catalog # MOLE41851510-1G
Marketsource
Supplier:   Molekula
γ-Cyclodextrine
Catalog # MOLE41851510-1G
Supplier:  Molekula
Marketsource
CAS Number:  17465-86-0
γ-Cyclodextrine

Glass bottle for solids, 1 g

CAS Number:  17465-86-0
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • EINECS:241-482-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications