Order Entry
Belgium
Orders LinkContactUsLinkComponent
γ-Cyclodextrine

γ-Cyclodextrine

γ-Cyclodextrine
Supplier:  MP Biomedicals
Certificates
γ-Cyclodextrine

3 Options Available Below

Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • EINECS:241-482-4

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 135,00 €
    135,00 €
    1 * 500 mg
       135,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 271,00 €
    271,00 €
    1 * 1 g
       271,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 620,00 €
    620,00 €
    1 * 5 g
       620,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.