Order Entry
Belgium
Orders LinkContactUsLinkComponent
β-Cyclodextrine

β-Cyclodextrine

Glass bottle for solids, 200 mg
Catalog # EDQMB0950000
Supplier:   EDQM
β-Cyclodextrine
Catalog # EDQMB0950000
Supplier:  EDQM
CAS Number:  7585-39-9
β-Cyclodextrine

Glass bottle for solids, 200 mg

CAS Number:  7585-39-9
Price
270,00 €
Log in for availability
1 * 200 mg
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.