Order Entry
Belgium
Orders LinkContactUsLinkComponent
α-Cyclodextrin

α-Cyclodextrin

Glass bottle for solids, 25 g
Catalog # MOLE15673368-25G
Marketsource
Supplier:   Molekula
α-Cyclodextrin
Catalog # MOLE15673368-25G
Supplier:  Molekula
Marketsource
CAS Number:  10016-20-3
α-Cyclodextrin

Glass bottle for solids, 25 g

CAS Number:  10016-20-3
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.