Sulphacetamide
Glass bottle for solids, 500 mg
Catalog # PROOMM0579.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: N-((4-Aminophenyl)sulphonyl)acetamide
- CAS Number:144-80-9
- Formula:C₈H₁₀N₂O₃S
- MDL Number:MFCD00066501
- Molecular Weight:214.24 g/mol
- EINECS:205-640-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



