Order Entry
Belgium
ContactUsLinkComponent
Sodium standard solution, 100 mg/L Na in water (from NaCl) standard for AAS
Sodium standard solution, 100 mg/L Na in water (from NaCl) standard for AAS
Catalog # KRAF03387.2000
Supplier:  Bernd Kraft
CAS Number:  
Sodium standard solution, 100 mg/L Na in water (from NaCl) standard for AAS
Catalog # KRAF03387.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item