Order Entry
Belgium
ContactUsLinkComponent
 
Sodium standard solution, 10 mg/L Na in water (from NaCl) standard for AAS
  KRAF03386.2000
 :  
Sodium standard solution, 10 mg/L Na in water (from NaCl) standard for AAS
  KRAF03386.2000
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Sodium
  • Symbol:
    Na