Molybdenum standard solution, 5000 mg/L Mo in 2 M nitric acid (from (NH₄)₆Mo₇O₂₄.4H₂O) standard for AAS
Catalog # KRAF03658.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:2 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Molybdenum
- Symbol:Mo
Specifications
About this item
Certifications: Traceable to NIST