VWR®, Glycylglycine (Gly-Gly)
0137-1KG
:
- Pack type:HDPE bottle
- Pk:1 kg
- CAS No.:556-50-3
- Seq:02
- Storage conditions:Ambient
- MDL No.:MFCD00008130
- Protein synonyms:Glycylglycine-Gly-Gly
- Protein/peptide name:Glycylglycine (Gly-Gly)
- Purity:>99%
- Molecular weight:132,12 g/mol
- Grade:High purity grade
- Molecular formula:C₄H₈N₂O₃
Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.
- Loss on drying: ≤0,2%
- Solubility (10%, water): Pass
- EINECS: 209-127-8
- GHS symbol and signal word: GHS07 and warning
Glycylglycine (Gly-Gly) contains ≤0,02% ammonia, ≤0,0001% arsenic, ≤0,2% ash, ≤0,02% chloride, ≤0,002% heavy metals (as Pb), ≤0,001% iron and ≤0,03% sulphate.