Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, 1,5-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
Pesticide reference standard, 1,5-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
Catalog # PROOCIL-ECN-2622
CAS Number:  
Pesticide reference standard, 1,5-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
Catalog # PROOCIL-ECN-2622
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    1825-30-5
  • Description:
    1,5-Dichloronaphthalene 100 µg/ml in n-nonane
  • Formula:
    C₁₀H₆Cl₂
  • Description:
    1,5-Dichloronaphthalene

Specifications

About this item