Silicon standard solution, 5000 mg/L Si in 3 M sodium hydroxide solution (from SiCl₄) standard for AAS
Catalog # KRAF03345.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:3 M NaOH
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Silicon
- Symbol:Si
Specifications
About this item
Certifications: Traceable to NIST